missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-Methylenemalonate (Stabilized with TBC, >85%), TRC

Product Code. 30385540
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385540 10 mg
30371135 50 mg
30373011 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385540 Supplier Toronto Research Chemicals Supplier No. TRCD49594510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3377-20-6
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
Synonym 1,3-Diethyl 2-Methylenepropanedioate, (Methylene)propanedioic Acid Diethyl Ester, 1,1-Dicarbethoxyethene, 1,3-Diethyl 2-methylidenepropanedioate, Diethyl Methylenemalonate, Ethyl Methylenemalonate
IUPAC Name diethyl 2-methylidenepropanedioate
SMILES CCOC(=O)C(=C)C(=O)OCC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H12O4/c1-4-11-7(9)6(3)8(10)12-5-2/h3-5H2,1-2H3
Formula Weight 172.0736
Chemical Name or Material Diethyl 2-Methylenemalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.