CAS RN 3377-20-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diethyl 2-Methylenemalonate (Stabilized with TBC, >85%), TRC
CAS: 3377-20-6 Molecular Formula: C8H12O4 Molecular Weight (g/mol): 172.18 Synonym: 1,3-Diethyl 2-Methylenepropanedioate,(Methylene)propanedioic Acid Diethyl Ester,1,1-Dicarbethoxyethene,1,3-Diethyl 2-methylidenepropanedioate,Diethyl Methylenemalonate,Ethyl Methylenemalonate IUPAC Name: diethyl 2-methylidenepropanedioate SMILES: CCOC(=O)C(=C)C(=O)OCC