missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 10-undecyn-1-ylphosphonate, 95%, Thermo Scientific™

Product Code. 15456057
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456057 250 mg 250mg
15446057 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456057 Supplier Thermo Scientific Alfa Aesar Supplier No. H33242.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1242248-76-5
Molecular Formula C15H29O3P
Molecular Weight (g/mol) 288.368
MDL Number MFCD17012685
InChI Key ZCOVBEAZCAHJDG-UHFFFAOYSA-N
Synonym diethyl 10-undecyn-1-ylphosphonate, diethyl undec-10-yn-1-ylphosphonate, 10-undecyn-1-ylphosphonic acid diethyl ester
PubChem CID 97638951
IUPAC Name 11-diethoxyphosphorylundec-1-yne
SMILES CCOP(=O)(CCCCCCCCCC#C)OCC

Specifications

Linear Formula HC≡C(CH2)9P(O)(OCH2CH3)2
Refractive Index 1.4460-1.4500
Quantity 250 mg
Formula Weight 288.37
Percent Purity 95%
Chemical Name or Material Diethyl 10-undecyn-1-ylphosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.