accessibility menu, dialog, popup

UserName

H3COPOCHOCH3

CAS RN 1242248-76-5

IUPAC Name: diethyl (undec-10-yn-1-yl)phosphonate
Synonyms: diethyl undec-10-yn-1-ylphosphonate
Molecular Weight (g/mol): 288.37
Molecular Formula: C15H29O3P
InChi Key: ZCOVBEAZCAHJDG-UHFFFAOYSA-N
SMILES: CCOP(=O)(CCCCCCCCCC#C)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results