missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ DIEA/NMP (N,N-diisopropylethylamine/N-methylpyrrolidone) (2 M)
Shop All Applied Biosystems Products
Click to view available options
:
Store at room temperature.
Description
- For Research Use Only
- Not for use in diagnostics procedures
Specifications
Specifications
| Format | Liquid |
| Product Type | N N-Diisopropylethylamine/N-Methylpyrrolidone |
| Content And Storage | Store at room temperature. |
| Reagent Type | Peptide Synthesis Reagent |
| Shipping Condition | Room Temperature |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion