missing translation for 'onlineSavingsMsg'
Learn More

Dideoxycytidinene, TRC

Product Code. 30383547
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383547 25 mg
30376056 1 mg
30370291 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383547 Supplier Toronto Research Chemicals Supplier No. TRCD43988025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7481-88-1
Molecular Formula C9 H11 N3 O3
Molecular Weight (g/mol) 209.2
Synonym Cytidine, 2',3'-didehydro-2',3'-dideoxy-, Cytosine, 1-(2,3-dideoxy-β-D-glycero-pent-2-enofuranosyl)- (8CI), 2',3'-Didehydro-2',3'-dideoxycytidine, 2',3'-Dideoxycytidin-2'-ene, 2'-Cytidinene, 2',3'-dideoxy-, D 4C, Dideoxycytidinene
IUPAC Name 4-amino-1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)C=C2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C9H11N3O3/c10-7-3-4-12(9(14)11-7)8-2-1-6(5-13)15-8/h1-4,6,8,13H,5H2,(H2,10,11,14)/t6-,8+/m0/s1
Formula Weight 209.08
Chemical Name or Material 2',3'-Didehydro-2',3'-dideoxycytidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.