CAS RN 7481-88-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dideoxycytidinene, TRC
CAS: 7481-88-1 Molecular Formula: C9 H11 N3 O3 Molecular Weight (g/mol): 209.2 Synonym: Cytidine, 2',3'-didehydro-2',3'-dideoxy-,Cytosine, 1-(2,3-dideoxy-β-D-glycero-pent-2-enofuranosyl)- (8CI),2',3'-Didehydro-2',3'-dideoxycytidine,2',3'-Dideoxycytidin-2'-ene,2'-Cytidinene, 2',3'-dideoxy-,D 4C,Dideoxycytidinene IUPAC Name: 4-amino-1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]pyrimidin-2-one SMILES: NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)C=C2