missing translation for 'onlineSavingsMsg'
Learn More

Didecylamine, TRC

Product Code. 30380612
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380612 1 g
30384194 10 g
30376058 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380612 Supplier Toronto Research Chemicals Supplier No. TRCD4394301G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-49-6
Molecular Formula C20 H43 N
Molecular Weight (g/mol) 297.56
Synonym N-Decyldecanamine, N-Decyl-1-decanaminde, Didecylamine (6CI,7CI,8CI), N,N-Didecylamine
IUPAC Name N-decyldecan-1-amine
SMILES CCCCCCCCCCNCCCCCCCCCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H43N/c1-3-5-7-9-11-13-15-17-19-21-20-18-16-14-12-10-8-6-4-2/h21H,3-20H2,1-2H3
Formula Weight 297.3396
Chemical Name or Material Didecylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.