accessibility menu, dialog, popup

UserName

H3CHNCH3

CAS RN 1120-49-6

IUPAC Name: bis(decyl)amine
Synonyms: bis(decyl)amine
Molecular Weight (g/mol): 297.57
Molecular Formula: C20H43N
InChi Key: GMTCPFCMAHMEMT-UHFFFAOYSA-N
SMILES: CCCCCCCCCCNCCCCCCCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results