missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac Epolamine, TRC

Product Code. 30380007
Change view
Click to view available options
Quantity:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380007 2.5 g
30385938 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380007 Supplier Toronto Research Chemicals Supplier No. TRCD4364702.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119623-66-4
Molecular Formula C14 H11 Cl2 N O2 . C6 H13 N O
Molecular Weight (g/mol) 411.32
Synonym Benzeneacetic acid, 2-[(2,6-dichlorophenyl)amino]-, compd. with 1-pyrrolidineethanol (1:1), 1-Pyrrolidineethanol, 2-[(2,6-dichlorophenyl)amino]benzeneacetate (salt) (9CI), Diclofenac 1-(2-hydroxyethyl)pyrrolidine salt, Diclofenac N-(2-hydroxyethyl)pyrrolidine salt, Diclofenac epolamine, Flector Patch
IUPAC Name 2-[2-(2,6-dichloroanilino)phenyl]acetic acid;2-pyrrolidin-1-ylethanol
SMILES OCCN1CCCC1.OC(=O)Cc2ccccc2Nc3c(Cl)cccc3Cl

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C14H11Cl2NO2.C6H13NO/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;8-6-5-7-3-1-2-4-7/h1-7,17H,8H2,(H,18,19);8H,1-6H2
Formula Weight 410.1164
Chemical Name or Material Diclofenac Epolamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.