CAS RN 119623-66-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diclofenac Epolamine, TRC
CAS: 119623-66-4 Molecular Formula: C14 H11 Cl2 N O2 . C6 H13 N O Molecular Weight (g/mol): 411.32 Synonym: Benzeneacetic acid, 2-[(2,6-dichlorophenyl)amino]-, compd. with 1-pyrrolidineethanol (1:1),1-Pyrrolidineethanol, 2-[(2,6-dichlorophenyl)amino]benzeneacetate (salt) (9CI),Diclofenac 1-(2-hydroxyethyl)pyrrolidine salt,Diclofenac N-(2-hydroxyethyl)pyrrolidine salt,Diclofenac epolamine,Flector Patch IUPAC Name: 2-[2-(2,6-dichloroanilino)phenyl]acetic acid;2-pyrrolidin-1-ylethanol SMILES: OCCN1CCCC1.OC(=O)Cc2ccccc2Nc3c(Cl)cccc3Cl