Learn More
Agilent BioTek Half-size Dichroic Mirror
Product Code. 12542787 Shop All Agilent BioTek Products
Click to view available options
Wavelength:
510 nm
545 nm
595 nm
Unit Size:
Each
Product Code. 12542787
Brand: Agilent BioTek 7138545
For use with Cytation 5, Synergy H1 and Synergy LX
For use with Cytation 5, Synergy H1 and Synergy LX
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dichroic Mirror
For Use With (Equipment) Cytation 3, Cytation 5, Synergy 2, Synergy H1, Ex. 512-535, Em. 555-578
Wavelength 545 nm