Learn More
Agilent BioTek Half-size Dichroic Mirror
For use with Cytation 5, Synergy H1 and Synergy LX
Brand:  Agilent BioTek 7138510
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12522787
  • £625.00 / Each
Estimated Shipment: 24-05-2024
to see stock.
Description
Description
For use with Cytation 5, Synergy H1 and Synergy LX
Specifications
Specifications
Dichroic Mirror
Cytation 3, Cytation 5, Synergy 2, Synergy H1, Ex. 440-505, Em. 515-640
510 nm