missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetic anhydride, 98%

Product Code. 10657522 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10657522 10 g Glass bottle 10g
10562951 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10657522 Supplier Thermo Scientific Acros Supplier No. 165120100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4124-30-5 | C4H2Cl4O3 | 239.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4124-30-5
Molecular Formula C4H2Cl4O3
Molecular Weight (g/mol) 239.87
MDL Number MFCD00000842
InChI Key RQHMQURGSQBBJY-UHFFFAOYSA-N
Synonym dichloroacetic anhydride, dichloroacetic acid anhydride, acetic acid, dichloro-, anhydride, unii-mb3ogr64in, anhydrid kyseliny dichloroctove, 2,2'-dichloroacetic anhydride, dichloroacetetic anhydride, mb3ogr64in, anhydrid kyseliny dichloroctove czech, acetic acid, 2,2-dichloro-, 1,1'-anhydride
PubChem CID 77772
IUPAC Name (2,2-dichloroacetyl) 2,2-dichloroacetate
SMILES C(C(=O)OC(=O)C(Cl)Cl)(Cl)Cl

Specifications

Melting Point 29.0°C
Color Colorless
Density 1.5700g/mL
Boiling Point 214.0°C to 216.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (Cl2CHCO)2O
Refractive Index 1.4825 to 1.4845
Quantity 10 g
Beilstein 02, 204
Specific Gravity 1.57
Solubility Information Solubility in water: decomposes in water. Other solubilities: decomposes in alcohol
Formula Weight 239.87
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Dichloroacetic anhydride, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dichloroacetic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.