accessibility menu, dialog, popup

UserName

ClClOOOClCl

CAS RN 4124-30-5

IUPAC Name: 2,2-dichloroacetyl 2,2-dichloroacetate
Synonyms: 2,2-dichloroacetyl 2,2-dichloroacetate
Molecular Weight (g/mol): 239.86
Molecular Formula: C4H2Cl4O3
InChi Key: RQHMQURGSQBBJY-UHFFFAOYSA-N
SMILES: ClC(Cl)C(=O)OC(=O)C(Cl)Cl
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)