missing translation for 'onlineSavingsMsg'
Learn More

Dichlobentiazox, TRC

Product Code. 30386474
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386474 250 mg -
30381066 25 mg 25mg
30368969 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386474 Supplier Toronto Research Chemicals Supplier No. TRCD434403250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957144-77-3
Molecular Formula C11 H6 Cl2 N2 O3 S2
Molecular Weight (g/mol) 349.21
Synonym 3-[(3,4-Dichloro-5-isothiazolyl)methoxy]-1,2-benzisothiazole 1,1-dioxide, Dichlobentiazox
IUPAC Name 3-[(3,4-dichloro-1,2-thiazol-5-yl)methoxy]-1,2-benzothiazole 1,1-dioxide
SMILES Clc1nsc(COC2=NS(=O)(=O)c3ccccc23)c1Cl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H6Cl2N2O3S2/c12-9-7(19-14-10(9)13)5-18-11-6-3-1-2-4-8(6)20(16,17)15-11/h1-4H,5H2
Formula Weight 347.9197
Chemical Name or Material Dichlobentiazox
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.