CAS RN 957144-77-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dichlobentiazox, TRC
CAS: 957144-77-3 Molecular Formula: C11 H6 Cl2 N2 O3 S2 Molecular Weight (g/mol): 349.21 Synonym: 3-[(3,4-Dichloro-5-isothiazolyl)methoxy]-1,2-benzisothiazole 1,1-dioxide,Dichlobentiazox IUPAC Name: 3-[(3,4-dichloro-1,2-thiazol-5-yl)methoxy]-1,2-benzothiazole 1,1-dioxide SMILES: Clc1nsc(COC2=NS(=O)(=O)c3ccccc23)c1Cl