missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicer Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-23891SS
This item is not returnable.
View return policy
Description
Dicer Polyclonal specifically detects Dicer in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications
| Dicer | |
| Polyclonal | |
| Flow Cytometry 1:100 - 1:1000, ELISA 1:200, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) reported in scientific literature (PMID 16823371) | |
| Q9UPY3 | |
| DICER1 | |
| Within the range of amino acids 1423-1463 of human Dicer protein were used as the immunogen for this antibody. | |
| 0.025 mL | |
| Apoptosis, Cell Biology, Cell Cycle and Replication, Chromatin Research, Cytokine Research, Epigenetics, Immunology, Innate Immunity, microRNA, Signal Transduction | |
| 23405 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Crude whole Rabbit sera, 50% Glycerol with 0.025% Sodium Azide | |
| DCR1, Dicer, dicer 1, double-stranded RNA-specific endoribonuclease, dicer 1, ribonuclease type III, Dicer1, Dcr-1 homolog (Drosophila), EC 3.1.26, EC 3.1.26.-, EC 3.1.26.3, Helicase MOI, Helicase with RNase motif, helicase-moi, HERNADicer1, Dcr-1 homolog, K12H4.8-LIKE, KIAA0928endoribonuclease Dicer | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction