missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicer Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£167.00 - £360.00
Specifications
| Antigen | Dicer |
|---|---|
| Dilution | Flow Cytometry 1:100 - 1:1000, ELISA 1:200, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) reported in scientific literature (PMID 16823371) |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18119601
|
Novus Biologicals
NBP2-23891SS |
0.025 mL |
£167.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18706943
|
Novus Biologicals
NBP2-23891 |
0.1 mL |
£360.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dicer Polyclonal specifically detects Dicer in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
| Dicer | |
| Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology, Cell Cycle and Replication, Chromatin Research, Cytokine Research, Epigenetics, Immunology, Innate Immunity, microRNA, Signal Transduction | |
| Crude whole Rabbit sera, 50% Glycerol with 0.025% Sodium Azide | |
| DCR1, Dicer, dicer 1, double-stranded RNA-specific endoribonuclease, dicer 1, ribonuclease type III, Dicer1, Dcr-1 homolog (Drosophila), EC 3.1.26, EC 3.1.26.-, EC 3.1.26.3, Helicase MOI, Helicase with RNase motif, helicase-moi, HERNADicer1, Dcr-1 homolog, K12H4.8-LIKE, KIAA0928endoribonuclease Dicer | |
| DICER1 | |
| IgG | |
| Unpurified |
| Flow Cytometry 1:100 - 1:1000, ELISA 1:200, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) reported in scientific literature (PMID 16823371) | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| Q9UPY3 | |
| 23405 | |
| Within the range of amino acids 1423-1463 of human Dicer protein were used as the immunogen for this antibody. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title