missing translation for 'onlineSavingsMsg'
Learn More

Dibucaine hydrochloride, Thermo Scientific™

Potent, long-acting local anesthetic

£40.80 - £134.00

Chemical Identifiers

CAS 61-12-1
Molecular Formula C20H30ClN3O2
Molecular Weight (g/mol) 379.929
MDL Number MFCD00012735
InChI Key IVHBBMHQKZBJEU-UHFFFAOYSA-N
Synonym dibucaine hydrochloride, dibucaine hcl, cinchocaine hydrochloride, cinchocaine hcl, cincaine chloride, dibucaine cinchocaine hcl, percain, percaine, sovcain
PubChem CID 521951
ChEBI CHEBI:59735
IUPAC Name 2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride
SMILES CCCCOC1=NC2=CC=CC=C2C(=C1)C(=O)NCCN(CC)CC.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15495099
SDS View Documents
Thermo Scientific Alfa Aesar
J62804.06
5g
£40.80
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15405109
SDS View Documents
Thermo Scientific Alfa Aesar
J62804.14
25g
£134.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is applied as a potent, long-acting local anesthetic. It is used as an anaesthetic agent for spinal anaesthesia. More commonly it is a component in rectal medications used in inflammation of the anus and for the symptomatic treatment of haemorrhoids and fissures in the anus.

Solubility
Freely soluble in water, alcohol, acetone, or chloroform

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

61-12-1
379.929
IVHBBMHQKZBJEU-UHFFFAOYSA-N
521951
2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride
C20H30ClN3O2
MFCD00012735
dibucaine hydrochloride, dibucaine hcl, cinchocaine hydrochloride, cinchocaine hcl, cincaine chloride, dibucaine cinchocaine hcl, percain, percaine, sovcain
CHEBI:59735
CCCCOC1=NC2=CC=CC=C2C(=C1)C(=O)NCCN(CC)CC.Cl

Specifications

61-12-1
MFCD00012735
dibucaine hydrochloride, dibucaine hcl, cinchocaine hydrochloride, cinchocaine hcl, cincaine chloride, dibucaine cinchocaine hcl, percain, percaine, sovcain
IVHBBMHQKZBJEU-UHFFFAOYSA-N
2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride
521951
379.92
C20H30ClN3O2
14,3031
Freely soluble in water, alcohol, acetone, or chloroform
CCCCOC1=NC2=CC=CC=C2C(=C1)C(=O)NCCN(CC)CC.Cl
379.929
CHEBI:59735
Dibucaine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

missing translation for 'einecsNumber' : 200-498-1

missing translation for 'rtecsNumber' : GD3325000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.