missing translation for 'onlineSavingsMsg'
Learn More

Dibucaine hydrochloride, Thermo Scientific™

Potent, long-acting local anesthetic

Brand:  Thermo Scientific Alfa Aesar J62804.14

Quantity 25g

 View more versions of this product

Product Code. 15405109

  • £134.00 / 25g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is applied as a potent, long-acting local anesthetic. It is used as an anaesthetic agent for spinal anaesthesia. More commonly it is a component in rectal medications used in inflammation of the anus and for the symptomatic treatment of haemorrhoids and fissures in the anus.

Solubility
Freely soluble in water, alcohol, acetone, or chloroform

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

61-12-1
379.929
IVHBBMHQKZBJEU-UHFFFAOYSA-N
521951
2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride
C20H30ClN3O2
MFCD00012735
dibucaine hydrochloride, dibucaine hcl, cinchocaine hydrochloride, cinchocaine hcl, cincaine chloride, dibucaine cinchocaine hcl, percain, percaine, sovcain
CHEBI:59735
CCCCOC1=NC2=CC=CC=C2C(=C1)C(=O)NCCN(CC)CC.Cl

Specifications

61-12-1
MFCD00012735
14,3031
Freely soluble in water, alcohol, acetone, or chloroform
CCCCOC1=NC2=CC=CC=C2C(=C1)C(=O)NCCN(CC)CC.Cl
379.929
CHEBI:59735
Dibucaine hydrochloride
C20H30ClN3O2
25g
dibucaine hydrochloride, dibucaine hcl, cinchocaine hydrochloride, cinchocaine hcl, cincaine chloride, dibucaine cinchocaine hcl, percain, percaine, sovcain
IVHBBMHQKZBJEU-UHFFFAOYSA-N
2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride
521951
379.92
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.