missing translation for 'onlineSavingsMsg'
Learn More

Dibromocyanoacetamide-13C3, TRC

Product Code. 30387697
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387697 1 mg
30377939 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387697 Supplier Toronto Research Chemicals Supplier No. TRCD4253121MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1286131-38-1
Molecular Formula 13C3H2Br2N2O
Molecular Weight (g/mol) 244.85
Synonym 2,2-Dibromo-2-cyanoacetamide-13C3, 2,2-Dibromo-3-nitrilopropanamide-13C3, Amerstat 300-13C3, BE 3S-13C3, Biomate 723-13C3, DBNPA-13C3, Dibromonitrilo- propionamide-13C3, Dowicil QK 20-13C3, Mucosin NT-13C3, NSC 98283-13C3, X-CIDE 508-13C3
SMILES [13C](#N)[13C]([13C](=O)N)(Br)Br

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C3H2Br2N2O/c4-3(5,1-6)2(7)8/h(H2,7,8)/i1+1,2+1,3+1
Chemical Name or Material Dibromocyanoacetamide-13C3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.