CAS RN 1286131-38-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dibromocyanoacetamide-13C3, TRC
CAS: 1286131-38-1 Molecular Formula: 13C3H2Br2N2O Molecular Weight (g/mol): 244.85 Synonym: 2,2-Dibromo-2-cyanoacetamide-13C3,2,2-Dibromo-3-nitrilopropanamide-13C3,Amerstat 300-13C3,BE 3S-13C3,Biomate 723-13C3,DBNPA-13C3,Dibromonitrilo- propionamide-13C3,Dowicil QK 20-13C3,Mucosin NT-13C3,NSC 98283-13C3,X-CIDE 508-13C3 SMILES: [13C](#N)[13C]([13C](=O)N)(Br)Br