missing translation for 'onlineSavingsMsg'
Learn More

Dibenzazepine (Deshydroxy LY 411575), TRC

Product Code. 30383652
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30383652 1 mg -
30372046 5 mg -
30371655 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383652 Supplier Toronto Research Chemicals Supplier No. TRCD4226001MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 209984-56-5
Molecular Formula C26 H23 F2 N3 O3
Molecular Weight (g/mol) 463.48
Synonym Dibenzazepine
IUPAC Name (2S)-2-[[2-(3,5-difluorophenyl)acetyl]amino]-N-[(7S)-5-methyl-6-oxo-7H-benzo[d][1]benzazepin-7-yl]propanamide
SMILES C[C@H](NC(=O)Cc1cc(F)cc(F)c1)C(=O)N[C@@H]2C(=O)N(C)c3ccccc3c4ccccc24

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C26H23F2N3O3/c1-15(29-23(32)13-16-11-17(27)14-18(28)12-16)25(33)30-24-21-9-4-3-7-19(21)20-8-5-6-10-22(20)31(2)26(24)34/h3-12,14-15,24H,13H2,1-2H3,(H,29,32)(H,30,33)/t15-,24-/m0/s1
Formula Weight 463.1707
Chemical Name or Material Dibenzazepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.