missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Diazoxide, 98%

Product Code. 15467199
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467199 500 mg 500mg
15457199 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467199 Supplier Thermo Scientific Chemicals Supplier No. J66010.ME
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 364-98-7 | C8H7ClN2O2S, C8H7ClN2O2S | 230.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 364-98-7

Specifications

Quantity 500 mg
Description White to off-white
Content And Storage 2°C to 8°C
Format Powder
Molecular Formula C8H7ClN2O2S
Formula Weight 230.67
Synonym 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide;
Melting Point 330°C to 331°C
Solubility Soluble in 0.1M NaOH. Insoluble in water or in methanol.
Form Powder
Assay 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.