missing translation for 'onlineSavingsMsg'
Learn More

Diallylmethylamine, 98%

Product Code. 15315078 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15315078 5 g 5g
15336713 25 g 25g
15325078 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15315078 Supplier Thermo Scientific Acros Supplier No. 456340050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2424-01-3 | C7H13N | 111.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2424-01-3
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.18

Specifications

Density 0.7890g/mL
Boiling Point 111.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Specific Gravity 0.789
Synonym 1,4-bis glycidyloxy benzene, hydroquinone diglycidyl ether, 2-4-oxiran-2-ylmethoxy phenoxymethyl oxirane, hydroquinone 1,4-diglycidyl ether, oxirane, 2,2'-1,4-phenylenebis oxymethylene bis, 2,2'-1,4-phenylenebis oxymethylene dioxirane, 2-4-oxiran-2-ylmethoxy phenoxy methyl oxirane, 1,4-bis oxiran-2-ylmethoxy benzene, acmc-20ap2z
InChI Key FSYPIGPPWAJCJG-UHFFFAOYNA-N
SMILES C1C(O1)COC2=CC=C(C=C2)OCC3CO3
IUPAC Name 2-[[4-(oxiran-2-ylmethoxy)phenoxy]methyl]oxirane
PubChem CID 62443
Formula Weight 111.18
Percent Purity 98%
Chemical Name or Material Diallylmethylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diallylmethylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.