accessibility menu, dialog, popup

UserName

CAS RN 2424-01-3

IUPAC Name: methylbis(prop-2-en-1-yl)amine
Synonyms: methylbis(prop-2-en-1-yl)amine
Molecular Weight (g/mol): 111.19
Molecular Formula: C7H13N
InChi Key: WGESLFUSXZBFQF-UHFFFAOYSA-N
SMILES: CN(CC=C)CC=C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results