Learn More
Di-n-octyldichlorotin, 95%, Thermo Scientific Chemicals
£48.70 - £344.00
Chemical Identifiers
CAS | 3542-36-7 |
---|---|
Molecular Formula | C16H36Cl2Sn |
Molecular Weight (g/mol) | 418.07 |
MDL Number | MFCD00018091 |
InChI Key | DXPJPPSKHFECLR-UHFFFAOYSA-N |
Synonym | dichlorodioctyltin, dioctyltin dichloride, di-n-octyltin dichloride, stannane, dichlorodioctyl, dotc, di-n-octyltindichloride, dichloro dioctyl stannane, di-n-octyldichlorotin, stannane, dioctyldichloro, di-n-octyl-zinn dichlorid |
PubChem CID | 77080 |
IUPAC Name | dichloro(dioctyl)stannane |
SMILES | Cl.Cl.CCCCCCCC[SnH2]CCCCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11378898
|
Thermo Scientific Alfa Aesar
041738.06 |
5 g |
£48.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358898
|
Thermo Scientific Alfa Aesar
041738.14 |
25 g |
£169.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368898
|
Thermo Scientific Alfa Aesar
041738.22 |
100 g | |||||||
Description
Di-n-octyldichlorotin compounds are used as intermediates in the production of other organotin chemicals. Metatin and Acima. These products are also used as catalysts or industrial intermediates in the production of polyurethanes, silicone resins, synthetic lubricants, and MS polymer sealants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3542-36-7 | |
418.07 | |
DXPJPPSKHFECLR-UHFFFAOYSA-N | |
77080 | |
Cl.Cl.CCCCCCCC[SnH2]CCCCCCCC |
C16H36Cl2Sn | |
MFCD00018091 | |
dichlorodioctyltin, dioctyltin dichloride, di-n-octyltin dichloride, stannane, dichlorodioctyl, dotc, di-n-octyltindichloride, dichloro dioctyl stannane, di-n-octyldichlorotin, stannane, dioctyldichloro, di-n-octyl-zinn dichlorid | |
dichloro(dioctyl)stannane |
Specifications
3542-36-7 | |
175°C (1 mmHg) | |
MFCD00018091 | |
Moisture sensitive | |
Insoluble in water. | |
Cl.Cl.CCCCCCCC[SnH2]CCCCCCCC | |
418.07 | |
416.04 | |
Crystalline |
44°C to 48°C | |
C16H36Cl2Sn | |
UN3261 | |
dichlorodioctyltin, dioctyltin dichloride, di-n-octyltin dichloride, stannane, dichlorodioctyl, dotc, di-n-octyltindichloride, dichloro dioctyl stannane, di-n-octyldichlorotin, stannane, dioctyldichloro, di-n-octyl-zinn dichlorid | |
DXPJPPSKHFECLR-UHFFFAOYSA-N | |
dichloro(dioctyl)stannane | |
77080 | |
95% | |
Di-n-octyldichlorotin |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 222-583-2
missing translation for 'rtecsNumber' : WH7247000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only