missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyldichlorotin, 95%

Product Code. 11368898
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368898 100 g 100g
11378898 5 g 5g
11358898 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368898 Supplier Thermo Scientific Alfa Aesar Supplier No. 041738.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3542-36-7 | C16H36Cl2Sn | 418.07 g/mol

Di-n-octyldichlorotin compounds are used as intermediates in the production of other organotin chemicals. Metatin and Acima. These products are also used as catalysts or industrial intermediates in the production of polyurethanes, silicone resins, synthetic lubricants, and MS polymer sealants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3542-36-7
Molecular Formula C16H36Cl2Sn
Molecular Weight (g/mol) 418.07
MDL Number MFCD00018091
InChI Key DXPJPPSKHFECLR-UHFFFAOYSA-N
Synonym dichlorodioctyltin, dioctyltin dichloride, di-n-octyltin dichloride, stannane, dichlorodioctyl, dotc, di-n-octyltindichloride, dichloro dioctyl stannane, di-n-octyldichlorotin, stannane, dioctyldichloro, di-n-octyl-zinn dichlorid
PubChem CID 77080
IUPAC Name dichloro(dioctyl)stannane
SMILES Cl.Cl.CCCCCCCC[SnH2]CCCCCCCC

Specifications

Melting Point 44°C to 48°C
Boiling Point 175°C (1 mmHg)
Quantity 100 g
UN Number UN3261
Sensitivity Moisture Sensitive
Formula Weight 416.04
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Di-n-octyldichlorotin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.