missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyl N,N-diethylcarbamoylphosphonate, 94%, Thermo Scientific™

Product Code. 11351597
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11351597 5 g 5g
11361597 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351597 Supplier Thermo Scientific Alfa Aesar Supplier No. L04480.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-69-2
Molecular Formula C13H28NO4P
Molecular Weight (g/mol) 293.344
MDL Number MFCD00039952
InChI Key VIZPJAFCJKGFQB-UHFFFAOYSA-N
Synonym di-n-butyl n,n-diethylcarbamoylphosphonate, dibutyl diethylamino carbonyl phosphonate, dibutyl diethylcarbamoylphosphonate, dibutoxycarbonyl-n,n-diethylcarboxamide, dibutyldiethylcarbamylphosphonate, dibutyl-n,n-diethylcar-bamylphosphonate, di-n-butyl n,n-diethylcarbamylphosphonate, n,n-diethylcarbamoylphosphonic acid dibutyl ester
PubChem CID 81944
IUPAC Name 1-dibutoxyphosphoryl-N,N-diethylformamide
SMILES CCCCOP(=O)(C(=O)N(CC)CC)OCCCC

Specifications

Boiling Point 132°C to 134°C (1 mmHg)
Refractive Index 1.453
Quantity 5 g
Beilstein 1796281
Formula Weight 293.34
Percent Purity 94%
Chemical Name or Material Di-n-butyl N,N-diethylcarbamoylphosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.