accessibility menu, dialog, popup

UserName

CH3OPOOCH3ONCH3H3C

CAS RN 7439-69-2

IUPAC Name: dibutyl (diethylcarbamoyl)phosphonate
Synonyms: dibutyl diethylcarbamoylphosphonate
Molecular Weight (g/mol): 293.34
Molecular Formula: C13H28NO4P
InChi Key: VIZPJAFCJKGFQB-UHFFFAOYSA-N
SMILES: CCCCOP(=O)(OCCCC)C(=O)N(CC)CC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results