missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyl maleate, 96%, Thermo Scientific™

Product Code. 15475338
Change view
Click to view available options
Quantity:
0.25 L
1 L
Unit Size:
0.25 L
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475338 1 L 1 L
15465338 0.25 L 0.25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475338 Supplier Thermo Scientific Alfa Aesar Supplier No. H59123.K2

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-76-0
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.29
MDL Number MFCD00009447,MFCD00065141
InChI Key JBSLOWBPDRZSMB-FPLPWBNLSA-N
Synonym dibutyl maleate, butyl maleate, staflex dbm, rc comonomer dbm, maleic acid, dibutyl ester, di-n-butyl maleate, 2-butenedioic acid z-, dibutyl ester, 2-butenedioic acid, dibutyl ester, maleic acid dibutyl ester, 2-butenedioic acid 2z-, dibutyl ester
PubChem CID 5271569
IUPAC Name dibutyl (Z)-but-2-enedioate
SMILES CCCCOC(=O)\C=C/C(=O)OCCCC

Specifications

Density 0.988
Boiling Point 281°C
Flash Point 138°C (280°F)
Linear Formula CH3(CH2)3OCOCH=CHCOO(CH2)3CH3
Refractive Index 1.445
Quantity 1 L
Beilstein 1726634
Formula Weight 228.28
Percent Purity 96%
Chemical Name or Material Di-n-butyl maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.