missing translation for 'onlineSavingsMsg'
Learn More

DHX58 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£134.00

Specifications

Antigen DHX58
Concentration 1 mg/mL
Dilution Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18541591
View Documents
Novus Biologicals
NBP1-76390-0.025mg
0.025 mg
£134.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DHX58 Polyclonal specifically detects DHX58 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

DHX58
Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000
Polyclonal
Rabbit
Human
EAW60798
79132
Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human LGP2. The immunogen is located within amino acids 90 - 140 of LGP2. Amino Acid Squence: ALTSPEEEEHVELT
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
1 mg/mL
Western Blot
Unconjugated
RUO
PBS with 0.02% Sodium Azide
D11lgp2e, D11LGP2probable ATP-dependent RNA helicase DHX58, DEXH (Asp-Glu-X-His) box polypeptide 58, EC 3.6.4.13, LGP2ortholog of mouse D11lgp2, Probable ATP-dependent helicase LGP2, Protein D11Lgp2 homolog, RIG-I-like receptor LGP2, RLR, RNA helicase LGP2
DHX58
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.