missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX58 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76390-0.025mg
This item is not returnable.
View return policy
Description
DHX58 Polyclonal specifically detects DHX58 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| DHX58 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| D11lgp2e, D11LGP2probable ATP-dependent RNA helicase DHX58, DEXH (Asp-Glu-X-His) box polypeptide 58, EC 3.6.4.13, LGP2ortholog of mouse D11lgp2, Probable ATP-dependent helicase LGP2, Protein D11Lgp2 homolog, RIG-I-like receptor LGP2, RLR, RNA helicase LGP2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79132 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000 | |
| EAW60798 | |
| DHX58 | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human LGP2. The immunogen is located within amino acids 90 - 140 of LGP2. Amino Acid Squence: ALTSPEEEEHVELT | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction