missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | DHRS9 |
|---|---|
| Concentration | 0.28 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16883254
|
Proteintech
14560-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16873254
|
Proteintech
14560-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DHRS9 is a 3-alpha-hydroxysteroid dehydrogenase that converts 3-alpha-tetrahydroprogesterone (allopregolone) to dihydroxyprogesterone and 3-alpha-androstanediol to dihydroxyprogesterone.Specifications
| DHRS9 | |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q58NB6, Q8VD48, Q9BPW9 | |
| 10170, 170635, 241452 | |
| DHRS9 Fusion Protein Ag6116 | |
| Primary | |
| -20°C | |
| DHRS9 |
| 0.28 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 3 alpha HSD, 3ALPHA HSD, DHRS9, RDH E2, RDH15, RDHL, RDHTBE, RETSDR8, SDR9C4 | |
| Dhrs9 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title