missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14560-1-AP-150UL
This item is not returnable.
View return policy
Description
DHRS9 is a 3-alpha-hydroxysteroid dehydrogenase that converts 3-alpha-tetrahydroprogesterone (allopregolone) to dihydroxyprogesterone and 3-alpha-androstanediol to dihydroxyprogesterone.Specifications
| DHRS9 | |
| Polyclonal | |
| Unconjugated | |
| DHRS9 | |
| 3 alpha HSD, 3ALPHA HSD, DHRS9, RDH E2, RDH15, RDHL, RDHTBE, RETSDR8, SDR9C4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10170, 170635, 241452 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q58NB6, Q8VD48, Q9BPW9 | |
| Dhrs9 | |
| DHRS9 Fusion Protein Ag6116 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction