missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGKQ Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17885-1-AP-150UL
This item is not returnable.
View return policy
Description
Diacylglycerol (DAG) is a fundamental lipid second messenger that is produced in the nucleus. The accumulation of DAG in the nucleus is important for the regulation of cell growth and differentiation. Diacylglycerol kinases (DGKs) convert DAG to phosphatidic acid, thereby terminating diacylglycerol signaling, which results in the reduction of protein kinase C activity and cell cycle progression of T lymphocytes. Diacylglycerol kinases are divided into five subtypes, Type I-Type V. DGK-theta is a Type V DGK, and localizes mainly to the nucleus of various cell lines, such as MDA-MB-453, MCF-7, PC12 and HeLa. Nuclear DGK-theta co-localizes with phosphatidylinositol 4, 5-bisphosphate (PIP(2)). DGK-theta is the isoform responsive to alpha-Thrombin stimulation.Specifications
| DGKQ | |
| Polyclonal | |
| Unconjugated | |
| DGKQ | |
| DAG kinase theta, DAGK, DAGK4, DAGK7, DGK theta, DGKQ, Diacylglycerol kinase theta, Diglyceride kinase theta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100361138, 110524, 1609 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52824, Q6P5E8 | |
| Dgkq | |
| DGKQ Fusion Protein Ag11938 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction