missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGCR8 Antibody (2H2), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | DGCR8 |
|---|---|
| Clone | 2H2 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043591
|
Novus Biologicals
NBP3-26623-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043902
|
Novus Biologicals
NBP3-26623-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DGCR8 Monoclonal antibody specifically detects DGCR8 in Human samples. It is validated for Western Blot, ELISASpecifications
| DGCR8 | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C22orf12, DGCRK6chromosome 22 open reading frame 12, DiGeorge syndrome critical region 8, DiGeorge syndrome critical region gene 8, Gy1, microprocessor complex subunit DGCR8, pasha | |
| A synthesized peptide derived from Human DGCR8 [UniProt Q8WYQ5] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 2H2 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Epigenetics, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 54487 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title