missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGCR8 Antibody (2H2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26623-50ul
This item is not returnable.
View return policy
Description
DGCR8 Monoclonal antibody specifically detects DGCR8 in Human samples. It is validated for Western Blot, ELISA
Specifications
| DGCR8 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 2H2 | |
| Western Blot 1:500-1:5000, ELISA | |
| C22orf12, DGCRK6chromosome 22 open reading frame 12, DiGeorge syndrome critical region 8, DiGeorge syndrome critical region gene 8, Gy1, microprocessor complex subunit DGCR8, pasha | |
| A synthesized peptide derived from Human DGCR8 [UniProt Q8WYQ5] | |
| 50 μL | |
| Epigenetics, Stem Cell Markers | |
| 54487 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction