missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DFNB31 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | DFNB31 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
18605391
|
Novus Biologicals
NBP2-92038-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623231
|
Novus Biologicals
NBP2-92038-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DFNB31 Polyclonal antibody specifically detects DFNB31 in Human, Mouse samples. It is validated for Western BlotSpécification
| DFNB31 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Vision | |
| PBS with 50% glycerol, pH7.3. | |
| 25861 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Autosomal recessive deafness type 31 protein, CIP98DKFZp434N014, deafness, autosomal recessive 31, DFNB 31, KIAA1526whirlin, USH2DWI, whrn, WHRNCASK-interacting protein CIP98 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 708-907 of human WHRN (NP_056219.3). PSGHPDQTGTNQHFVMVEVHRPDSEPDVNEVRALPQTRTASTLSQLSDSGQTLSEDSGVDAGEAEASAPGRGRQSVSTKSRSSKELPRNERPTDGANKPPGLLEPTSTLVRVKKSAATLGIAIEGGANTRQPLPRIVTIQRGGSAHNCGQLKVGHVILEVNGLTLRGKEHREAARIIAEAFKTKDRDYIDFLVTEFNVML | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit