missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DFNB31 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92038-0.02ml
This item is not returnable.
View return policy
Description
DFNB31 Polyclonal antibody specifically detects DFNB31 in Human, Mouse samples. It is validated for Western Blot
Specifications
| DFNB31 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Autosomal recessive deafness type 31 protein, CIP98DKFZp434N014, deafness, autosomal recessive 31, DFNB 31, KIAA1526whirlin, USH2DWI, whrn, WHRNCASK-interacting protein CIP98 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 708-907 of human WHRN (NP_056219.3). PSGHPDQTGTNQHFVMVEVHRPDSEPDVNEVRALPQTRTASTLSQLSDSGQTLSEDSGVDAGEAEASAPGRGRQSVSTKSRSSKELPRNERPTDGANKPPGLLEPTSTLVRVKKSAATLGIAIEGGANTRQPLPRIVTIQRGGSAHNCGQLKVGHVILEVNGLTLRGKEHREAARIIAEAFKTKDRDYIDFLVTEFNVML | |
| 0.02 mL | |
| Vision | |
| 25861 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction