missing translation for 'onlineSavingsMsg'
Learn More

Dextran (Technical Grade~ 10K), TRC

Product Code. 30006404
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006404 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30006404 Supplier Toronto Research Chemicals Supplier No. TRCD29911010G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9004-54-0
Molecular Formula H2(C6H9O5)n(C7H12O5)mOH
Quantity 10 g
Synonym Dextrans, DEX 500, Detrax 40, Dextranen, Dextraven, Eudextran, Expandex, G 75, Gentran, Gentran 40, Hemodex, Hyscon, Hyskon, Infucoll, Intrader, Intradex, LMD, LMWD, LU 122, LVD, Longasteril 70, Macrodex, Macrose, Oncovertin N, Onkotin, PL 1S, Plasmafusin, Plasmodex, Plavolex, Polyglucin, Polyglusol, Promit, Rheodextran, Rheoisodex, Rheomacrodex, Rheopolyglucin, Rheopolyglucine, Rheorondex, Rheotran, Rondex, Serva G, T 10, T 110, T 150, T 20, T 250, T 4, T 40, T 5, T 500, T 6, T 70, Ultradex, α-Dextran, Dextran 1.5, Dextran 10, Dextran 1000, Dextran 10000, Dextran 110, Dextran 15, Dextran 150, Dextran 2000, Dextran 20000, Dextran 250, Dextran 3000, Dextran 40, Dextran 40000, Dextran 45, Dextran 500, Dextran 60, Dextran 70, Dextran 75, Dextran B 512, Dextran B1355, Dextran D 10, Dextran PL 1S, Dextran PT 25, Dextran PVD, Dextran RMI, Dextran T 10, Dextran T 100, Dextran T 110, Dextran T 150, Dextran T 20, Dextran T 2000, Dextran T 40, Dextran T 500, Dextran T 70;
InChI Formula InChI=1S/C17H32O10/c1-4-8-12(18)16(22)13(19)10(26-8)6-25-7-11-15(21)17(24-3)14(20)9(27-11)5-23-2/h8-22H,4-7H2,1-3H3/t8-,9-,10-,11-,12+,13-,14-,15+,16-,17+/m1/s1
SMILES O[C@@H]1[C@@H](COC)O[C@H](COC[C@@H]2[C@@H](O)[C@H](O)[C@@H](O)[C@@H](CC)O2)[C@H](O)[C@H]1OC
IUPAC Name (2R,3S,4R,5R,6R)-2-((((2R,3S,4S,5R,6R)-3,5-dihydroxy-4-methoxy-6-(methoxymethyl)tetrahydro-2H-pyran-2-yl)methoxy)methyl)-6-ethyltetrahydro-2H-pyran-3,4,5-triol
Molecular Weight (g/mol) 504.4
Chemical Name or Material Dextran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.