Learn More
Dexamethasone 21-phosphate disodium salt, 98%, Thermo Scientific™
Dexamethasone 21-phosphate disodium salt, CAS # 2392-39-4, is a synthetic adrenal glucocorticoid with potent anti-inflammatory activity.
£85.65 - £293.00
Chemical Identifiers
| CAS | 2392-39-4 |
|---|---|
| Molecular Formula | C22H28FNa2O8P |
| Molecular Weight (g/mol) | 516.41 |
| MDL Number | MFCD00079105 |
| InChI Key | PLCQGRYPOISRTQ-FCJDYXGNSA-L |
| Synonym | dexamethasone sodium phosphate, dexamethasone 21-phosphate disodium salt, dalalone, dexadreson, dexagro, megacort, soldesam, spersadox, decdan, solu-decadron |
| PubChem CID | 16961 |
| ChEBI | CHEBI:4462 |
| IUPAC Name | disodium;[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate |
| SMILES | CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COP(=O)([O-])[O-])O)C)O)F)C.[Na+].[Na+] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15466189
|
Thermo Scientific Alfa Aesar
J64083.03 |
1g |
£95.40 £85.65 / 1g Save £9.75 10% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15476189
|
Thermo Scientific Alfa Aesar
J64083.06 |
5g |
£293.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 2392-39-4 | |
| 516.41 | |
| PLCQGRYPOISRTQ-FCJDYXGNSA-L | |
| 16961 | |
| disodium;[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate |
| C22H28FNa2O8P | |
| MFCD00079105 | |
| dexamethasone sodium phosphate, dexamethasone 21-phosphate disodium salt, dalalone, dexadreson, dexagro, megacort, soldesam, spersadox, decdan, solu-decadron | |
| CHEBI:4462 | |
| CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COP(=O)([O-])[O-])O)C)O)F)C.[Na+].[Na+] |
Specifications
| 2392-39-4 | |
| MFCD00079105 | |
| 142943 | |
| Soluble in water. Slightly soluble in ethanol. Insoluble in dichloromethane. | |
| CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COP(=O)([O-])[O-])O)C)O)F)C.[Na+].[Na+] | |
| 516.41 | |
| CHEBI:4462 | |
| 98% |
| C22H28FNa2O8P | |
| 6473066 | |
| dexamethasone sodium phosphate, dexamethasone 21-phosphate disodium salt, dalalone, dexadreson, dexagro, megacort, soldesam, spersadox, decdan, solu-decadron | |
| PLCQGRYPOISRTQ-FCJDYXGNSA-L | |
| disodium;[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate | |
| 16961 | |
| 516.4 | |
| Dexamethasone 21-phosphate disodium salt |
Safety and Handling
Harmful if swallowed, Suspected of damaging the unborn child, May cause damage to organs through prolonged or repeated exposure
Acute toxicity (category 4), Reproductive toxicity (category 2), Specific target organ toxicity after repeated exposure (category 2)
missing translation for 'einecsNumber' : 219-243-0
missing translation for 'rtecsNumber' : TU4056000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.