missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone 21-phosphate disodium salt, 98%

Product Code. 15476189
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476189 5 g 5g
15466189 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476189 Supplier Thermo Scientific Alfa Aesar Supplier No. J64083.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dexamethasone 21-phosphate disodium salt, CAS # 2392-39-4, is a synthetic adrenal glucocorticoid with potent anti-inflammatory activity. | CAS: 2392-39-4 | C22H28FNa2O8P | 516.41 g/mol

Dexamethasone 21-phosphate disodium salt acts as an inducer of apoptosis and inhibitor of the sodium phosphate symporter. It is used in the treatment of emetic effects of cancer therapy and epicondylitis. Further, it serves as an activator of food, medicine, glucocorticoid and anti inflammatory.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Dexamethasone 21-phosphate disodium salt is a sodium phosphate salt form of dexamethasone, considered to be a synthetic adrenal glucocorticoid with anti-inflammatory activity

Application

  • Dexamethasone 21-phosphate disodium salt binds to specific nuclear steroid receptors, interfering with NF-kB activation and apoptotic pathways
  • It acts as an inducer of apoptosis and inhibitor of the sodium phosphate symporter
  • It has been used as a positive control to study its effects on ocular shedding of herpes simplex virus type 1 in the induced reactivation and spontaneous shedding rabbit models
  • This compound is used to study its protective effects against cisplatin-induced ototoxicity in the inner ear of guinea pigs
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2392-39-4
Molecular Formula C22H28FNa2O8P
Molecular Weight (g/mol) 516.41
MDL Number MFCD00079105
InChI Key PLCQGRYPOISRTQ-FCJDYXGNSA-L
Synonym dexamethasone sodium phosphate, dexamethasone 21-phosphate disodium salt, dalalone, dexadreson, dexagro, megacort, soldesam, spersadox, decdan, solu-decadron
PubChem CID 16961
ChEBI CHEBI:4462
IUPAC Name disodium;[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate
SMILES CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COP(=O)([O-])[O-])O)C)O)F)C.[Na+].[Na+]

Specifications

Melting Point 233°C to 240°C
Color White
Quantity 5 g
Beilstein 6473066
Merck Index 142943
Solubility Information Soluble in water. Slightly soluble in ethanol. Insoluble in dichloromethane.
Formula Weight 516.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Dexamethasone 21-phosphate disodium salt
compliance-icons compliance-icons
Product Identifier
  • Dexamethasone 21-phosphate disodium salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Dexamethasone 21-phosphate disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.