Learn More
Fisherbrand™ Device Cover for Elmasonic X-tra ST Ultrasonic Cleaners
Product Code. 15636084 Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H/1600 H
Ultrasonic Cleaner, Elmasonic Xtra ST 1900S
Ultrasonic Cleaner, Elmasonic Xtra ST 2500 H
Ultrasonic Cleaner, Elmasonic Xtra ST 600 H/800 H
Ultrasonic Cleaner, Elmastonic Xtra ST 300 H/500 H
Unit Size:
Each
Product Code. 15636084
Only null left
Add to basket
Add to basket
Provides a secure closure for ultrasonic cleaning baths.
  • Flat design with handle
  • Available in a range of sizes for all X-tra ST models
  • Manufactured from stainless steel
  • Accessory item
Specifications
Type Lid
For Use With (Equipment) Ultrasonic Cleaner, Elmasonic Xtra ST 2500 H
For Use With (Application) Closure for X-tra ST Ultrasonic Cleaners
Includes Handle
Shape Flat