missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Destrin Recombinant Rabbit Monoclonal Antibody (23GB5225), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Destrin |
|---|---|
| Clone | 23GB5225 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089078
![]() |
Invitrogen™
MA552818 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088760
|
Invitrogen™
MA552819 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
Destrin Recombinant Monoclonal antibody specifically detects Destrin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifica
| Destrin | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P60981, Q7M0E3, Q9R0P5 | |
| 11034, 502674, 56431 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5225 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2610043P17Rik; ACTDP; actin depolymerizing factor; actin-depolymerizing factor; ADF; AU042046; bA462D18.2; bA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)); corn1; corneal disease 1; destrin; destrin (actin depolymerizing factor); destrin, actin depolymerizing factor; Dsn; DSTN; epididymis luminal protein 32; HEL32; RP23-77A2.1; sID; Sid 23; Sid23; sid23p | |
| A synthesized peptide derived from human Destrin (50-100AA). | |
| Primary | |
| -20°C | |
| DSTN |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto