missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Destrin Recombinant Rabbit Monoclonal Antibody (23GB5225), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552819
This item is not returnable.
View return policy
Description
The product of this gene belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This gene encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin). Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| Destrin | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DSTN | |
| 2610043P17Rik; ACTDP; actin depolymerizing factor; actin-depolymerizing factor; ADF; AU042046; bA462D18.2; bA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)); corn1; corneal disease 1; destrin; destrin (actin depolymerizing factor); destrin, actin depolymerizing factor; Dsn; DSTN; epididymis luminal protein 32; HEL32; RP23-77A2.1; sID; Sid 23; Sid23; sid23p | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11034, 502674, 56431 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5225 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P60981, Q7M0E3, Q9R0P5 | |
| DSTN | |
| A synthesized peptide derived from human Destrin (50-100AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction