missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoplakin Antibody (SR1430), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £358.00
Specifications
| Antigen | Desmoplakin |
|---|---|
| Clone | SR1430 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684175
|
Novus Biologicals
NBP3-22092-100ul |
100 μg |
£358.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600204
|
Novus Biologicals
NBP3-22092-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Desmoplakin Monoclonal antibody specifically detects Desmoplakin in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
| Desmoplakin | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 1832 | |
| IgG | |
| Affinity purified |
| SR1430 | |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| 250/210 kDa paraneoplastic pemphigus antigen, desmoplakin, desmoplakin (DPI, DPII), desmoplakin I, desmoplakin II, DP, DPI, DPII, KPPS2, PPKS2 | |
| A synthesized peptide derived from human Desmoplakin (Uniprot #: P15924) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title