missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoplakin Antibody (SR1430), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22092-50ul
This item is not returnable.
View return policy
Description
Desmoplakin Monoclonal antibody specifically detects Desmoplakin in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Specifications
| Desmoplakin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1832 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| SR1430 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| 250/210 kDa paraneoplastic pemphigus antigen, desmoplakin, desmoplakin (DPI, DPII), desmoplakin I, desmoplakin II, DP, DPI, DPII, KPPS2, PPKS2 | |
| A synthesized peptide derived from human Desmoplakin (Uniprot #: P15924) | |
| 50 μg | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction