missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Desmoglein 2 Recombinant Rabbit Monoclonal Antibody (23GB5215), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553867
This item is not returnable.
View return policy
Description
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
Specifications
| Desmoglein 2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DSG2 | |
| AA408168; ARVC10; ARVD10; Cadherin family member 5; CDHF5; CMD1BB; D18Ertd293e; desmoglein 2; desmoglein-2; DSG2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1829 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5215 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14126 | |
| DSG2 | |
| A synthesized peptide derived from human Desmoglein 2 (1050-1118AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction